siRNA Details
Gene Symbol: 1700018G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045422.1 | 3 | 306 | |||
GenBank mRNA | AK006087.1 | - | 3 | 307 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706200.1 | - | 3 | 307 | Chr. X - 102928372 - 102929094 [+] on Build GRCm38 |
