siRNA Details
| Gene Symbol: 1700018G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045422.1 | 3 | 248 | |||
| GenBank mRNA | AK006087.1 | - | 3 | 249 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706200.1 | - | 3 | 249 | Chr. X - 102928372 - 102929094 [+] on Build GRCm38 | |

1700018G05Rik
Genomic Map