siRNA Details
| Gene Symbol: 2700089I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045308.1 | 4 | 436 | |||
| GenBank mRNA | AK012591.1 | - | 4 | 437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700089I24Rik| Gene Symbol: 2700089I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045308.1 | 4 | 436 | |||
| GenBank mRNA | AK012591.1 | - | 4 | 437 | ||