siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012593.1 | - | 1 | 248 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711738.1 | - | 1 | 245 | Chr. 15 - 80884353 - 80884727 [+] on Build GRCm38 | |

Tnrc6b
Genomic Map