siRNA Details
| Gene Symbol: 9130208D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA790037.1 | - | 1 | 328 | ||
| AK020274.1 | - | 1 | 349 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720846.1 | - | 1 | 349 | Chr. 7 - 105823251 - 105823806 [-] on Build GRCm38 | |

9130208D14Rik
Genomic Map