siRNA Details
| Gene Symbol: 9130208D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA790037.1 | - | 1 | 240 | ||
| AK020274.1 | - | 1 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130208D14Rik| Gene Symbol: 9130208D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA790037.1 | - | 1 | 240 | ||
| AK020274.1 | - | 1 | 261 | |||