siRNA Details
| Gene Symbol: 9130209A04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033453.1 | 4 | 416 | |||
| GenBank mRNA | AK020276.1 | - | 4 | 417 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY720848.1 | - | 4 | 417 | Chr. 18 - 47452668 - 47470855 [+] on Build GRCm38 | |

9130209A04Rik
Genomic Map