siRNA Details
| Gene Symbol: 1700064F23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006880.1 | - | 2 | 331 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706866.1 | - | 2 | 331 | Chr. 8 - 110006880 - 110007654 [+] on Build GRCm38 | |

1700064F23Rik
Genomic Map