siRNA Details
| Gene Symbol: 1700064M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045288.1 | 2 | 565 | |||
| GenBank mRNA | AK006884.1 | - | 2 | 566 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706870.1 | - | 2 | 566 | Chr. 12 - 99626075 - 99627974 [-] on Build GRCm38 | |

1700064M15Rik
Genomic Map