siRNA Details
| Gene Symbol: 1700064M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045288.1 | 1 | 298 | |||
| GenBank mRNA | AK006884.1 | - | 1 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700064M15Rik| Gene Symbol: 1700064M15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045288.1 | 1 | 298 | |||
| GenBank mRNA | AK006884.1 | - | 1 | 299 | ||