siRNA Details
| Gene Symbol: BB166591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037728.1 | - | 1 | 3479 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CV675857.1 | - | 1 | 698 | Chr. 2 - 152824362 - 152825098 [-] on Build GRCm38 | |

BB166591
Genomic Map