siRNA Details
Gene Symbol: BB166591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA672916.1 | - | 1 | 499 | ||
AK037728.1 | - | 1 | 4553 | |||
BB166591.1 | - | 1 | 312 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BB166591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA672916.1 | - | 1 | 499 | ||
AK037728.1 | - | 1 | 4553 | |||
BB166591.1 | - | 1 | 312 |