siRNA Details
| Gene Symbol: 1700040P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006664.1 | - | 1 | 138 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN843523.1 | - | 123 | Chr. - - [-] on Build | ||

1700040P09Rik
Genomic Map