siRNA Details
| Gene Symbol: Bcas1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040610.1 | 7, 8 | 921 | |||
| GenBank mRNA | AK006663.1 | - | 7, 8 | 921 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcas1os2| Gene Symbol: Bcas1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040610.1 | 7, 8 | 921 | |||
| GenBank mRNA | AK006663.1 | - | 7, 8 | 921 | ||