siRNA Details
| Gene Symbol: Bcas1os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040610.1 | 5 | 507 | |||
| GenBank mRNA | AK006663.1 | - | 5 | 507 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706651.1 | - | 5 | 503 | Chr. 2 - 170356365 - 170378758 [+] on Build GRCm38 | |

Bcas1os2
Genomic Map