siRNA Details
| Gene Symbol: 1700040K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006660.1 | - | 1 | 170 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706647.1 | - | 1 | 170 | Chr. 1 - 152936604 - 152937043 [+] on Build GRCm38 | |

1700040K01Rik
Genomic Map