siRNA Details
| Gene Symbol: 1110035E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004108.1 | - | 1 | 379 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY704351.1 | - | 1 | 379 | Chr. 8 - 107312677 - 107313671 [+] on Build GRCm38 | |

1110035E04Rik
Genomic Map