siRNA Details
| Gene Symbol: A930012N16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020850.1 | - | 1 | 213 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY742182.1 | - | 1 | 194 | Chr. 2 - 156180595 - 156181113 [+] on Build GRCm38 | |

A930012N16Rik
Genomic Map