siRNA Details
| Gene Symbol: 4933433G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040719.1 | 2 | 670 | |||
| GenBank mRNA | AK014947.1 | - | 2 | 672 | ||
| AK017040.1 | - | 671 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714464.1 | - | 2 | 666 | Chr. 9 - 75410188 - 75414377 [+] on Build GRCm38 | |

4933433G15Rik
Genomic Map