siRNA Details
| Gene Symbol: Gm31121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK139864.1 | - | 5 | 439 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725998.1 | - | 5 | 436 | Chr. 3 - 149563639 - 149660393 [-] on Build GRCm38 | |

Gm31121
Genomic Map