siRNA Details
| Gene Symbol: Gm20740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK041852.1 | - | 1 | 4064 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB233142.2 | - | 1 | 574 | Chr. 15 - 63445126 - 63445730 [-] on Build GRCm38 | |

Gm20740
Genomic Map