siRNA Details
| Gene Symbol: Gm15169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK163197.1 | - | 2323 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB258363.2 | - | 1 | 747 | Chr. X - 157757427 - 157758173 [-] on Build GRCm38 | |

Gm15169
Genomic Map