siRNA Details
| Gene Symbol: Gm20740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045279.1 | 1 | 21 | |||
| GenBank mRNA | AK040104.1 | - | 1 | 22 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20740| Gene Symbol: Gm20740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045279.1 | 1 | 21 | |||
| GenBank mRNA | AK040104.1 | - | 1 | 22 | ||