siRNA Details
| Gene Symbol: A630089N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043155.1 | - | 740 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630089N07Rik| Gene Symbol: A630089N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043155.1 | - | 740 | |||