siRNA Details
| Gene Symbol: A630089N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043155.1 | - | 428 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB640464.1 | - | 4 | 425 | Chr. 16 - 98001451 - 98081990 [-] on Build GRCm38 | |

A630089N07Rik
Genomic Map