siRNA Details
| Gene Symbol: 9030407P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018490.1 | - | 2 | 1363 | ||
| AK034016.1 | - | 3 | 1572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030407P20Rik| Gene Symbol: 9030407P20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018490.1 | - | 2 | 1363 | ||
| AK034016.1 | - | 3 | 1572 | |||