siRNA Details
| Gene Symbol: Gm6307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045331.1 | 4 | 1165 | |||
| GenBank mRNA | AK140896.1 | - | 4 | 1168 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX729663.1 | - | 3 | 408 | Chr. 2 - 180386203 - 180401144 [+] on Build GRCm38 | |

Gm6307
Genomic Map