siRNA Details
| Gene Symbol: Gm6307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045331.1 | 4 | 1469 | |||
| GenBank mRNA | AK140896.1 | - | 4 | 1472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6307| Gene Symbol: Gm6307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045331.1 | 4 | 1469 | |||
| GenBank mRNA | AK140896.1 | - | 4 | 1472 | ||