siRNA Details
| Gene Symbol: Gm32461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085480.1 | - | 1 | 877 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB501117.2 | - | 1 | 82 | Chr. 16 - 86770286 - 86805572 [-] on Build GRCm38 | |

Gm32461
Genomic Map