siRNA Details
| Gene Symbol: Gm34045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136051.1 | - | 4 | 281 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719988.1 | - | 4 | 281 | Chr. 10 - 119135468 - 119166385 [-] on Build GRCm38 | |

Gm34045
Genomic Map