siRNA Details
| Gene Symbol: Gm34045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136051.1 | - | 6 | 822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm34045| Gene Symbol: Gm34045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136051.1 | - | 6 | 822 | ||