siRNA Details
| Gene Symbol: 4933424L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016895.1 | - | 1 | 219 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716575.1 | - | 1 | 215 | Chr. 12 - 106158780 - 106159730 [+] on Build GRCm38 | |

4933424L07Rik
Genomic Map