siRNA Details
Gene Symbol: Dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012530.1 | - | 715 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ746458.1 | - | 1 | 48 | Chr. 8 - 34821251 - 34821952 [+] on Build GRCm38 |
