siRNA Details
| Gene Symbol: 1700093E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007049.1 | - | 1 | 637 | ||
| AK018923.1 | - | 1 | 637 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707119.1 | - | 1 | 634 | Chr. 2 - 178417473 - 178418240 [-] on Build GRCm38 | |

1700093E11Rik
Genomic Map