siRNA Details
Gene Symbol: Gm26705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103487.1 | 3 | 272 | |||
GenBank mRNA | BC050000.1 | - | 3 | 272 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ929997.1 | - | 271 | Chr. - - [-] on Build |
