siRNA Details
| Gene Symbol: Gm26705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103487.1 | 4 | 446 | |||
| GenBank mRNA | BC050000.1 | - | 4 | 446 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ929997.1 | - | 445 | Chr. - - [-] on Build | ||

Gm26705
Genomic Map