siRNA Details
| Gene Symbol: Gm13497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040636.1 | 2 | 436 | |||
| GenBank mRNA | AK042686.1 | - | 2 | 437 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB639851.1 | - | 2 | 437 | Chr. 2 - 51241831 - 51294472 [+] on Build GRCm38 | |

Gm13497
Genomic Map