siRNA Details
| Gene Symbol: 4930552N02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040661.1 | 3, 4 | 292 | |||
| GenBank mRNA | AK016098.1 | - | 3, 4 | 293 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715700.1 | - | 3, 4 | 293 | Chr. 4 - 52814604 - 52827972 [+] on Build GRCm38 | |

4930552N02Rik
Genomic Map