siRNA Details
Gene Symbol: 4930552N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040661.1 | 4 | 1004 | |||
GenBank mRNA | AK016098.1 | - | 4 | 1005 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930552N02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040661.1 | 4 | 1004 | |||
GenBank mRNA | AK016098.1 | - | 4 | 1005 |