siRNA Details
| Gene Symbol: Gm32548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042350.1 | - | 2 | 243 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB639252.1 | - | 2 | 243 | Chr. 18 - 73711326 - 73717202 [+] on Build GRCm38 | |

Gm32548
Genomic Map