siRNA Details
| Gene Symbol: 1700027F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040681.1 | 3 | 712 | |||
| GenBank mRNA | AK006413.1 | - | 3 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700027F09Rik| Gene Symbol: 1700027F09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040681.1 | 3 | 712 | |||
| GenBank mRNA | AK006413.1 | - | 3 | 712 | ||