siRNA Details
| Gene Symbol: 1700027H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040594.1 | 2 | 78 | |||
| GenBank mRNA | AK006417.1 | - | 2 | 79 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706472.1 | - | 2 | 76 | Chr. 3 - 45416584 - 45439307 [-] on Build GRCm38 | |

1700027H10Rik
Genomic Map