siRNA Details
| Gene Symbol: Gm9054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045872.1 | 3 | 605 | |||
| GenBank mRNA | AK028005.1 | - | 3 | 605 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV018108.2 | - | 3 | 618 | Chr. 3 - 95985510 - 95986964 [+] on Build GRCm38 | |

Gm9054
Genomic Map