siRNA Details
| Gene Symbol: Fcnaos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039020.1 | - | 9 | 1320 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB180689.2 | - | 7 | 544 | Chr. 2 - 25625268 - 25631758 [+] on Build GRCm38 | |

Fcnaos
Genomic Map