siRNA Details
| Gene Symbol: 5031415H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017142.1 | - | 1 | 800 | ||
| AK081553.1 | - | 1 | 1473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5031415H12Rik| Gene Symbol: 5031415H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017142.1 | - | 1 | 800 | ||
| AK081553.1 | - | 1 | 1473 | |||