siRNA Details
| Gene Symbol: 5033403F01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017147.1 | - | 2 | 666 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716870.1 | - | 2 | 661 | Chr. 13 - 38960623 - 38965884 [+] on Build GRCm38 | |

5033403F01Rik
Genomic Map