siRNA Details
| Gene Symbol: 9030625N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018580.1 | - | 1 | 563 | ||
| AK035942.1 | - | 1 | 1130 | |||
| AK076245.1 | - | 1 | 1133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9030625N01Rik| Gene Symbol: 9030625N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018580.1 | - | 1 | 563 | ||
| AK035942.1 | - | 1 | 1130 | |||
| AK076245.1 | - | 1 | 1133 | |||