siRNA Details
| Gene Symbol: 9130001E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018581.1 | - | 220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130001E16Rik| Gene Symbol: 9130001E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018581.1 | - | 220 | |||