siRNA Details
| Gene Symbol: 2610318M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019187.1 | - | 1 | 205 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY711242.1 | - | 1 | 199 | Chr. 14 - 8368313 - 8368567 [+] on Build GRCm38 | |

2610318M16Rik
Genomic Map