siRNA Details
Gene Symbol: 9130401L11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036628.1 | - | 1 | 3868 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB137637.2 | - | 1 | 466 | Chr. 6 - 98967702 - 98968392 [-] on Build GRCm38 |
